Anti-ANO1 Mouse Monoclonal Antibody [clone: SPM580]
Toimittaja: Abnova
ABNOMAB12069EA 518 EUR
ABNOMAB12069
Anti-ANO1 Mouse Monoclonal Antibody [clone: SPM580]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant ANO1.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: ANO1
Clonality: Monoclonal
Clone: SPM580
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now