Anti-CD1A Mouse Monoclonal Antibody [clone: C1A/711]
Toimittaja: Abnova
ABNOMAB12124EA 518 EUR
ABNOMAB12124
Anti-CD1A Mouse Monoclonal Antibody [clone: C1A/711]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant human CD1A.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: C1A/711
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now