Anti-IGHG1 Mouse Monoclonal Antibody [clone: IG266]

Toimittaja: Abnova
ABNOMAB12129EA 518 EUR
ABNOMAB12129
Anti-IGHG1 Mouse Monoclonal Antibody [clone: IG266]
Vasta-aineet
Mouse monoclonal antibody raised against human IGHG1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IGHG1
Clonality: Monoclonal
Clone: IG266
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR