Anti-IGHM Mouse Monoclonal Antibody [clone: DA4-4]

Toimittaja: Abnova
ABNOMAB12130EA 518 EUR
ABNOMAB12130
Anti-IGHM Mouse Monoclonal Antibody [clone: DA4-4]
Vasta-aineet
Mouse monoclonal antibody raised against human IGHM.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: IGHM
Clonality: Monoclonal
Clone: DA4-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR