Anti-c-Myc tag Chicken Polyclonal Antibody (Biotin)

Toimittaja: Abnova
ABNOPAB29204EA 338 EUR
ABNOPAB29204
Anti-c-Myc tag Chicken Polyclonal Antibody (Biotin)
Vasta-aineet
Chicken polyclonal antibody raised against c-Myc tag with Biotin conjugation.

Recommended Dilutions: ELISA: 1:2500-1:10000; Western Blot: 1:500-1:1500

Type: Tag
Antigen: c-Myc tag
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR