Anti-CORT Mouse Monoclonal Antibody [clone: 8G10]

Toimittaja: Abnova
ABNOH00001325-M01EA 380 EUR
ABNOH00001325-M01
Anti-CORT Mouse Monoclonal Antibody [clone: 8G10]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant CORT.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CORT
Clonality: Monoclonal
Clone: 8G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR