Anti-CYP7A1 Mouse Monoclonal Antibody [clone: 8F1]

Toimittaja: Abnova
ABNOH00001581-M01EA 394 EUR
ABNOH00001581-M01
Anti-CYP7A1 Mouse Monoclonal Antibody [clone: 8F1]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant CYP7A1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: CYP7A1
Clonality: Monoclonal
Clone: 8F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR