Anti-ECT2 Mouse Monoclonal Antibody [clone: 5D4]
Toimittaja: Abnova
ABNOH00001894-M01EA 394 EUR
ABNOH00001894-M01
Anti-ECT2 Mouse Monoclonal Antibody [clone: 5D4]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant ECT2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: ECT2
Clonality: Monoclonal
Clone: 5D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now