Anti-SLC17A7 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S28-9]

Toimittaja: Abnova
ABNOMAB11525EA 643 EUR
ABNOMAB11525
Anti-SLC17A7 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S28-9]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant of Slc17a7.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A7
Clonality: Monoclonal
Clone: S28-9
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR