Anti-SLC17A6 Mouse Monoclonal Antibody (Biotin) [clone: S29-29]

Toimittaja: Abnova
ABNOMAB11541EA 772 EUR
ABNOMAB11541
Anti-SLC17A6 Mouse Monoclonal Antibody (Biotin) [clone: S29-29]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant of Slc17a6.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A6
Clonality: Monoclonal
Clone: S29-29
Conjugation: Biotin
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR