Anti-CACNA1G Mouse Monoclonal Antibody (ATTO 594) [clone: S178A-9]

Toimittaja: Abnova
ABNOMAB11552EA 545 EUR
ABNOMAB11552
Anti-CACNA1G Mouse Monoclonal Antibody (ATTO 594) [clone: S178A-9]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant of Cacna1g.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: ATTO 594
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR