Anti-CACNA1G Mouse Monoclonal Antibody (Biotin) [clone: S178A-9]
Toimittaja: Abnova
ABNOMAB11559EA 772 EUR
ABNOMAB11559
Anti-CACNA1G Mouse Monoclonal Antibody (Biotin) [clone: S178A-9]
Vasta-aineet
Mouse monoclonal antibody raised against recombinant of Cacna1g.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: Biotin
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now