Anti-ITGAL Mouse Monoclonal Antibody [clone: CRIS-3]

Toimittaja: Abnova
ABNOMAB12133EA 471 EUR
ABNOMAB12133
Anti-ITGAL Mouse Monoclonal Antibody [clone: CRIS-3]
Vasta-aineet
Mouse monoclonal antibody raised against human ITGAL.

Recommended Dilutions: Flow Cytometry Immunohistochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: CRIS-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR