Anti-CK8 Mouse Monoclonal Antibody [clone: TS1]
Toimittaja: Abnova
ABNOMAB12135EA 518 EUR
ABNOMAB12135
Anti-CK8 Mouse Monoclonal Antibody [clone: TS1]
Vasta-aineet
Mouse monoclonal antibody raised against human KRT8.
Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: CK8
Clonality: Monoclonal
Clone: TS1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now