Anti-Biotin Monoclonal Antibody [clone: Hyb-8]

Toimittaja: Abnova
ABNOMAB12148EA 440 EUR
ABNOMAB12148
Anti-Biotin Monoclonal Antibody [clone: Hyb-8]
Vasta-aineet
Mouse monoclonal antibody raised against biotin.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: Hyb-8
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR