Anti-GSTT1 Mouse Monoclonal Antibody [clone: S1]

Toimittaja: Abnova
ABNOH00002952-M01AEA 394 EUR
ABNOH00002952-M01A
Anti-GSTT1 Mouse Monoclonal Antibody [clone: S1]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant GSTT1.

Type: Primary
Antigen: GSTT1
Clonality: Monoclonal
Clone: S1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR