Anti-KCNB1 Mouse Monoclonal Antibody (ATTO 594) [clone: S89-34]

Toimittaja: Abnova
ABNOMAB11570EA 545 EUR
ABNOMAB11570
Anti-KCNB1 Mouse Monoclonal Antibody (ATTO 594) [clone: S89-34]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of Kcnb1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: KCNB1
Clonality: Monoclonal
Clone: S89-34
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR