Anti-CK19 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28668EA 545 EUR
ABNOPAB28668
Anti-CK19 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant KRT19.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: CK19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR