Anti-NME4 Mouse Monoclonal Antibody [clone: 4B6]

Toimittaja: Abnova
ABNOH00004833-M02EA 380 EUR
ABNOH00004833-M02
Anti-NME4 Mouse Monoclonal Antibody [clone: 4B6]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant NME4.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: NME4
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR