Anti-PPY Mouse Polyclonal Antibody
Toimittaja: Abnova
ABNOH00005539-B02EA 380 EUR
ABNOH00005539-B02
Anti-PPY Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human PPY protein.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: PPY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now