Anti-PICK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S20-8]

Toimittaja: Abnova
ABNOMAB11630EA 545 EUR
ABNOMAB11630
Anti-PICK1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: S20-8]
Vasta-aineet
Mouse monoclonal antibody raised against partial synthetic protein of Pick1.

Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PICK1
Clonality: Monoclonal
Clone: S20-8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR