Anti-GTPBP1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28703EA 518 EUR
ABNOPAB28703
Anti-GTPBP1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant GTPBP1.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: GTPBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR