Anti-MED15 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28707EA 518 EUR
ABNOPAB28707
Anti-MED15 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant MED15.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Immunofluorescence: 1-4 ?g/ml; Western Blot: 1:100-1:500

Type: Primary
Antigen: MED15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR