Anti-NDRG2 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28730EA 518 EUR
ABNOPAB28730
Anti-NDRG2 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant NDRG2.

Recommended Dilutions: Immunohistochemistry: 1:2500-1:5000; Immunofluorescence: 1-4 ?g/ml; Western Blot: 1:100-1:250

Type: Primary
Antigen: NDRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR