Anti-PLXNB2 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28733EA 518 EUR
ABNOPAB28733
Anti-PLXNB2 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant PLXNB2.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: PLXNB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR