Anti-GPHN Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB28735EA 518 EUR
ABNOPAB28735
Anti-GPHN Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant GPHN.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50; Western Blot: 1:100 - 1:250

Type: Primary
Antigen: GPHN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR