Anti-PTPN14 Mouse Monoclonal Antibody [clone: 4F7]
Toimittaja: Abnova
ABNOH00005784-M01EA 394 EUR
ABNOH00005784-M01
Anti-PTPN14 Mouse Monoclonal Antibody [clone: 4F7]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant PTPN14.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: PTPN14
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now