Anti-SCD Mouse Monoclonal Antibody [clone: 1B7]

Toimittaja: Abnova
ABNOH00006319-M02EA 394 EUR
ABNOH00006319-M02
Anti-SCD Mouse Monoclonal Antibody [clone: 1B7]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant SCD.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SCD
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR