Anti-CCL17 Mouse Monoclonal Antibody [clone: 2E7]

Toimittaja: Abnova
ABNOH00006361-M07AEA 394 EUR
ABNOH00006361-M07A
Anti-CCL17 Mouse Monoclonal Antibody [clone: 2E7]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant CCL17.

Type: Primary
Antigen: CCL17
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR