Anti-GRIA2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S21-32]
Toimittaja: Abnova
ABNOMAB11653EA 545 EUR
ABNOMAB11653
Anti-GRIA2 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: S21-32]
Vasta-aineet
Mouse monoclonal antibody raised against partial synthetic protein of Gria2.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: GRIA2
Clonality: Monoclonal
Clone: S21-32
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now