Anti-KCNA3 Mouse Monoclonal Antibody (ATTO 594) [clone: S23-27]
Toimittaja: Abnova
ABNOMAB11660EA 545 EUR
ABNOMAB11660
Anti-KCNA3 Mouse Monoclonal Antibody (ATTO 594) [clone: S23-27]
Vasta-aineet
Mouse monoclonal antibody raised against synthetic peptide of Kcna3.
Recommended Dilutions: Immunohistochemistry: 0.1-1 ?g/ml; Immunofluorescence: 1-10 ?g/ml; Western Blot: 1 ?g/ml
Type: Primary
Antigen: KCNA3
Clonality: Monoclonal
Clone: S23-27
Conjugation: ATTO 594
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now