Anti-SLC12A3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Toimittaja: Abnova
ABNOPAB28764EA 545 EUR
ABNOPAB28764
Anti-SLC12A3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Vasta-aineet
Rabbit polyclonal antibody raised against recombinant Human, Rat SLC12A3.
Recommended Dilutions: Immunofluorescence Western Blot: 1:1000
Type:
Antigen: SLC12A3
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now