Anti-LY6D Mouse Polyclonal Antibody
Toimittaja: Abnova
ABNOH00008581-B03EA 394 EUR
ABNOH00008581-B03
Anti-LY6D Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human LY6D protein.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: LY6D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now