Anti-IgG Chicken Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29185EA 399 EUR
ABNOPAB29185
Anti-IgG Chicken Polyclonal Antibody
Vasta-aineet
Chicken polyclonal antibody raised against native Human IgG F(ab')2.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR