Anti-OXSR1 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29214EA 419 EUR
ABNOPAB29214
Anti-OXSR1 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human OXSR1.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; Western Blot: 1:500-1000

Type: Primary
Antigen: OXSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR