Anti-MGC50273 Mouse Monoclonal Antibody [clone: 3B10]
Toimittaja: Abnova
ABNOH00408029-M01EA 394 EUR
ABNOH00408029-M01
Anti-MGC50273 Mouse Monoclonal Antibody [clone: 3B10]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant MGC50273.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: MGC50273
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now