Anti-TCEAL3 Mouse Monoclonal Antibody [clone: 4F3]

Toimittaja: Abnova
ABNOH00085012-M05EA 394 EUR
ABNOH00085012-M05
Anti-TCEAL3 Mouse Monoclonal Antibody [clone: 4F3]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant TCEAL3.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: TCEAL3
Clonality: Monoclonal
Clone: 4F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR