Anti-KLF10 Mouse Monoclonal Antibody [clone: 3A10]

Toimittaja: Abnova
ABNOH00007071-M47EA 394 EUR
ABNOH00007071-M47
Anti-KLF10 Mouse Monoclonal Antibody [clone: 3A10]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant KLF10.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: KLF10
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR