Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 2C8]
Toimittaja: Abnova
ABNOH00030008-M19EA 394 EUR
ABNOH00030008-M19
Anti-EFEMP2 Mouse Monoclonal Antibody [clone: 2C8]
Vasta-aineet
Mouse monoclonal antibody raised against a partial recombinant EFEMP2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: EFEMP2
Clonality: Monoclonal
Clone: 2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now