Anti-CMTM5 Mouse Polyclonal Antibody
Toimittaja: Abnova
ABNOH00116173-B01PEA 394 EUR
ABNOH00116173-B01P
Anti-CMTM5 Mouse Polyclonal Antibody
Vasta-aineet
Mouse polyclonal antibody raised against a full-length human CMTM5 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: CMTM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now