Anti-RARRES2 Mouse Monoclonal Antibody [clone: 3B4]
Toimittaja: Abnova
ABNOH00005919-M23EA 394 EUR
ABNOH00005919-M23
Anti-RARRES2 Mouse Monoclonal Antibody [clone: 3B4]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant RARRES2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: RARRES2
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now