Anti-KAT5 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29252EA 356 EUR
ABNOPAB29252
Anti-KAT5 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human KAT5.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100

Type: Primary
Antigen: KAT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR