Anti-C1orf31 Mouse Monoclonal Antibody [clone: 4A6]

Toimittaja: Abnova
ABNOH00388753-M03EA 394 EUR
ABNOH00388753-M03
Anti-C1orf31 Mouse Monoclonal Antibody [clone: 4A6]
Vasta-aineet
Mouse monoclonal antibody raised against a full-length recombinant C1orf31.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: C1orf31
Clonality: Monoclonal
Clone: 4A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR