Anti-EPHA3 Rabbit Polyclonal Antibody

Toimittaja: Abnova
ABNOPAB29297EA 356 EUR
ABNOPAB29297
Anti-EPHA3 Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human EPHA3/EPHA4/EPHA5.

Recommended Dilutions: Immunohistochemistry: 1:50~1:100

Type: Primary
Antigen: EPHA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR