Incubating shaker, WIS-30
Toimittaja: Witeg
WITGDH.WIS00230EA 7210 EUR
WITGDH.WIS00230
Incubating shaker, WIS-30
Ravistelijat Inkubaatioravistelija
For applications in biological cultures at various temperatures up to 60 °C, this shaking incubator accommodates flasks up to 1000 ml. The front door and sliding platform enable easy loading and handling of samples. The unit features an advanced shaking mechanism, providing quiet shaking with precise speed control and temperature control by means of a digital fuzzy controller.
  • Digital back-lit LCD display
  • Universal spring rack for various shapes/size of flask available as an option
  • Automatic stop of shaking, heating and refrigeration when a door opens
  • RS232 interface
  • Timer: 99 h 59 min or continuous
Toimitustiedot: Supplied with WiseRemote® remote control software and cable. A range of options and accessories are available, please enquire for details.
Varoitus: Accommodates a maximum of 35×50/100, 16×200/300 or 12×500/1000 ml flasks.