Multi-tube racks
Toimittaja: Ratiolab
RATI3712101EA 22.7 EUR
RATI3712101
Multi-tube racks
Polyamide (PA).
  • Innovative design of internal clamp fixes the test tubes centric and vertical at any time
  • Dishwasher safe
  • Max. working temperature 90 °C
Ideal for several microcentrifuge tubes, cryo tubes and test tubes with different diameters.