Anti-Peroxiredoxin-1 Rabbit Polyclonal Antibody
Toimittaja: Biosensis
BSENR-1590-100EA 312 EUR
BSENR-1590-100
Anti-Peroxiredoxin-1 Rabbit Polyclonal Antibody
Vasta-aineet
Peroxiredoxin-1 has a role in redox regulation of the cell.
Human and rat Peroxiredoxin 1
Application Information:
IHC, WB, ELISA. This antibody works in IHC on frozen or wax embedded tissues. Antigen retrieval has been used in testing but may not be necessary. Typical working dilutions for light microscopy are 1:500 to 1:1,000 and IF of 1:50 to 1:100. For WB a dilution range of 1: 1,000 to 1: 4,000 is recommended. A dilution of 1: 1,000 to 1: 4,000 is recommended for ELISA. This antibody stains non-ciliated bronchiolar cells in rat airways and specific kidney tubule cells. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: Peroxiredoxin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 103-116 amino acids of Human Peroxiredoxin-1
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat,