Anti-NT3 Sheep Polyclonal Antibody
Toimittaja: Biosensis
BSENS-053-100EA 302 EUR
BSENS-053-100
Anti-NT3 Sheep Polyclonal Antibody
Vasta-aineet
NT3 is a member of the neurotrophin family, that controls survival and differentiation of visceral and proprioceptive sensory neurons. NT3 is closely related to both NGF and BDNF. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NT3-deficient mice generated by gene targeting display sevvere movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Brain and peripheral tissues. SIMILARITY: Belongs to the NGF-beta family.
A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Application Information:
IHC, ELISA, WB, inhibition of biological activity. A dilution of 1:500 to 1:2000 is recommended for IHC, western blot. For inhibition of biological activity: 1:10-50 for in vitro, 5-10 µl/g body weight for in vivo. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: central region of NT3
Host: Sheep
Isotype: IgG
Reactivity: Rat, Chicken, Human