Anti-Connexin-45 Sheep Polyclonal Antibody

Toimittaja: Biosensis
BSENS-061-100EA 312 EUR
BSENS-061-100
Anti-Connexin-45 Sheep Polyclonal Antibody
Vasta-aineet
Connexin-45 is a component of gap junctions, which are composed of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low molecular weight diffuse from one cell to a neighboring cell. SUBUNIT: A connexon is composed of a hexamer of connexins. SUBCELLULAR LOCATION: Membrane; multi-pass membrane protein. SIMILARITY: Belongs to the connexin family. Alpha-type (group II) subfamily. Alternatively spliced isoforms have been described.

Western blotting of cell membranes of COS cells transiently transfected with cDNA encoding Connexin-45 shows a singular band of molecular weight 47 kDa which is abolished by preincubation with the immunising peptide. No such band is detected in western blots of membranes from non-transfected cells or cells transfected with cDNA encoding Connexin-37 or Connexin-40.
Application Information:
Immunofluorescence, Western blot. A dilution of 1:10000 to 1: 20000 is recommended for Immunofluorescence and 1:20000 to 1:50000 for western blot. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: Connexin-45
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human Connexin-45 protein (aa: 354-367)
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR