Anti-C-reactive Protein Sheep Polyclonal Antibody

Toimittaja: Biosensis
BSENS-071-100EA 312 EUR
BSENS-071-100
Anti-C-reactive Protein Sheep Polyclonal Antibody
Vasta-aineet
C-reactive protein has several roles associated with host defence such as; promoting agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. It can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. COFACTOR: Binds 2 calcium ions per subunit. C-reactive protein exists as a homopentamer. Pentaxin (or Pentraxin) have a discoid arrangement of 5 non-covalently bound subunits. There are 2 alternatively spliced isoforms. C-reactive protein is found in plasma and its concentration increases greatly during acute phase response to tissue injury, infection or other inflammatory stimuli. It is induced by IL-1 and IL-6.

It has been shown to be specific for C-reactive protein by WB.
Application Information:
WB. Extremely high titre antibody. For western blot a dilution of 1: 10,000 to 1: 100000 is recommended. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C-reactive Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR